Assay Details
| Gene Symbol: AEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096639.4 | XP_001096639.4 | 891 | 107 | ||
| XM_015151239.2 | XP_015006725.2 | 1083 | 107 | |||
| XM_015151240.2 | XP_015006726.1 | 528 | 107 | |||
| GenBank mRNA | JU327712.1 | - | 375 | 107 | ||
| JU327713.1 | - | 375 | 107 | |||
| JV641907.1 | - | 886 | 107 | |||
More Information
Set Membership: |
|

AEBP2
Genomic Map