Assay Details
| Gene Symbol: FAIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151683.2 | XP_015007169.2 | 743 | 64 | ||
| GenBank mRNA | JU326169.1 | - | 399 | 64 | ||
| JU474781.1 | - | 570 | 64 | |||
| JV527791.1 | - | 622 | 64 | |||
| JV709998.1 | - | 70 | 64 | |||
More Information
Set Membership: |
|

FAIM2
Genomic Map