Assay Details
| Gene Symbol: CCPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028850939.1 | XP_028706772.1 | 1225 | 88 | ||
| XM_028850940.1 | XP_028706773.1 | 1416 | 88 | |||
| XM_028850941.1 | XP_028706774.1 | 1028 | 88 | |||
| XM_028850942.1 | XP_028706775.1 | 1129 | 88 | |||
| XM_028850943.1 | XP_028706776.1 | 1329 | 88 | |||
| XM_028850944.1 | XP_028706777.1 | 929 | 88 | |||
| XM_028850945.1 | XP_028706778.1 | 1225 | 88 | |||
| XM_028850946.1 | XP_028706779.1 | 1225 | 88 | |||
| XM_028850947.1 | XP_028706780.1 | 1225 | 88 | |||
| XM_028850948.1 | XP_028706781.1 | 1129 | 88 | |||
| XM_028850949.1 | XP_028706782.1 | 1225 | 88 | |||
| GenBank mRNA | JU322935.1 | - | 974 | 88 | ||
| JU477838.1 | - | 913 | 88 | |||
| JV048695.1 | - | 905 | 88 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCPG1
Genomic Map