Assay Details
| Gene Symbol: CADM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799800.3 | XP_002799846.1 | 1439 | 79 | ||
| XM_002799801.3 | XP_002799847.1 | 1355 | 79 | |||
| XM_002799802.3 | XP_002799848.1 | 1352 | 79 | |||
| XM_002799803.3 | XP_002799849.1 | 1268 | 79 | |||
| XM_015115760.2 | XP_014971246.1 | 1385 | 79 | |||
| XM_015115761.2 | XP_014971247.1 | 1301 | 79 | |||
| GenBank mRNA | JU327907.1 | - | 1071 | 79 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CADM1
Genomic Map