Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142294.2 | XP_014997780.1 | 880 | 72 | ||
| XM_015142295.2 | XP_014997781.1 | 880 | 72 | |||
| XM_028850951.1 | XP_028706784.1 | 880 | 72 | |||
More Information
Set Membership: |
|

DNAAF4
Genomic Map