Assay Details
| Gene Symbol: CRYBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131966.2 | XP_014987452.2 | 4736 | 74 | ||
| XM_015131967.2 | XP_014987453.2 | 4646 | 74 | |||
| XM_028837611.1 | XP_028693444.1 | 3694 | 74 | |||
| XM_028837619.1 | XP_028693452.1 | 3718 | 74 | |||
| XM_028837623.1 | XP_028693456.1 | 3695 | 74 | |||
More Information
Set Membership: |
|

CRYBG2
Genomic Map