Assay Details
| Gene Symbol: PMS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133250.2 | XP_014988736.2 | 2644 | 89 | ||
| XM_015133251.2 | XP_014988737.2 | 2488 | 89 | |||
| XM_028845123.1 | XP_028700956.1 | 2543 | 89 | |||
| XM_028845124.1 | XP_028700957.1 | 2634 | 89 | |||
| XM_028845125.1 | XP_028700958.1 | 2552 | 89 | |||
| XM_028845126.1 | XP_028700959.1 | 2316 | 89 | |||
| GenBank mRNA | JU325424.1 | - | 2417 | 89 | ||
| JV717116.1 | - | 2516 | 89 | |||
More Information
Set Membership: |
|

PMS2
Genomic Map