Assay Details
| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266830.1 | NP_001253759.1 | 903 | 73 | ||
| GenBank mRNA | JU474464.1 | - | 951 | 73 | ||
| JV635980.1 | - | 903 | 73 | |||
More Information
Set Membership: |
|

RDH10
Genomic Map