Assay Details
| Gene Symbol: OTUD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015129797.2 | XP_014985283.1 | 513 | 94 | ||
| XM_015129802.2 | XP_014985288.1 | 514 | 94 | |||
| GenBank mRNA | JU474873.1 | - | 486 | 94 | ||
| JU474874.1 | - | 486 | 94 | |||
More Information
Set Membership: |
|

OTUD3
Genomic Map