Assay Details
| Gene Symbol: CMTM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097523.4 | XP_001097523.1 | 730 | 80 | ||
| GenBank mRNA | JU326855.1 | - | 351 | 80 | ||
| JU481119.1 | - | 108 | 80 | |||
More Information
Set Membership: |
|

CMTM8
Genomic Map