Assay Details
| Gene Symbol: IGFBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260547.2 | NP_001247476.1 | 884 | 66 | ||
| XM_015133508.2 | XP_014988994.1 | 880 | 66 | |||
| GenBank mRNA | JV043702.1 | - | 888 | 66 | ||
| JV635732.1 | - | 858 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IGFBP3
Genomic Map