Assay Details
| Gene Symbol: UFM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257925.2 | NP_001244854.1 | 266 | 72 | ||
| XM_028837096.1 | XP_028692929.1 | 367 | 72 | |||
| XM_028837097.1 | XP_028692930.1 | 325 | 72 | |||
| GenBank mRNA | CO774469.1 | - | 205 | 72 | ||
| JU320298.1 | - | 268 | 72 | |||
| JU472548.1 | - | 234 | 72 | |||
| JU472549.1 | - | 234 | 72 | |||
| JV047327.1 | - | 236 | 72 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UFM1
Genomic Map