Assay Details
| Gene Symbol: PRMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839900.1 | XP_028695733.1 | 160 | 59 | ||
| XM_028839902.1 | XP_028695735.1 | 259 | 59 | |||
| XM_028839903.1 | XP_028695736.1 | 74 | 59 | |||
| GenBank mRNA | JU328004.1 | - | 30 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028839902.1, XM_028839903.1 transcripts.
Set Membership: |
|

PRMT1
Genomic Map