Assay Details
| Gene Symbol: SCAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260562.2 | NP_001247491.1 | 212 | 66 | ||
| XM_015142707.2 | XP_014998193.1 | 486 | 66 | |||
| GenBank mRNA | JV043839.1 | - | 216 | 66 | ||
| JV669631.1 | - | 314 | 66 | |||
More Information
Set Membership: |
|

SCAMP5
Genomic Map