Assay Details
| Gene Symbol: SYT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114405.2 | XP_014969891.2 | 1097 | 72 | ||
| XM_015114406.2 | XP_014969892.1 | 853 | 72 | |||
| XM_028833490.1 | XP_028689323.1 | 817 | 72 | |||
| GenBank mRNA | JU335648.1 | - | 1025 | 72 | ||
| JV588818.1 | - | 1039 | 72 | |||
| JV691344.1 | - | 1065 | 72 | |||
More Information
Set Membership: |
|

SYT13
Genomic Map