Assay Details
| Gene Symbol: MRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261086.1 | NP_001248015.1 | 322 | 92 | ||
| GenBank mRNA | JU332756.1 | - | 322 | 92 | ||
| JU332757.1 | - | 322 | 92 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRAP2
Genomic Map