Assay Details
| Gene Symbol: CTCFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089797.4 | XP_001089797.2 | 1256 | 98 | ||
| XM_015148727.2 | XP_015004213.2 | 1256 | 98 | |||
| XM_015148729.2 | XP_015004215.2 | 451 | 98 | |||
| XM_015148730.2 | XP_015004216.2 | 674 | 98 | |||
| GenBank mRNA | JV052235.1 | - | 211 | 98 | ||
More Information
Set Membership: |
|

CTCFL
Genomic Map