Assay Details
| Gene Symbol: DYNLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119163.2 | XP_014974649.1 | 413 | 95 | ||
| GenBank mRNA | JU319616.1 | - | 382 | 95 | ||
| JU470950.1 | - | 383 | 95 | |||
| JV043379.1 | - | 383 | 95 | |||
More Information
Set Membership: |
|

DYNLL2
Genomic Map