Assay Details
| Gene Symbol: MPLKIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133679.2 | XP_014989165.1 | 664 | 84 | ||
| GenBank mRNA | JU332980.1 | - | 406 | 84 | ||
| JU474350.1 | - | 406 | 84 | |||
| JV639022.1 | - | 408 | 84 | |||
More Information
Set Membership: |
|

MPLKIP
Genomic Map