Assay Details
| Gene Symbol: ALDH5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101142.4 | XP_001101142.1 | 667 | 67 | ||
| GenBank mRNA | JV481605.1 | - | 152 | 67 | ||
| JV593391.1 | - | 470 | 67 | |||
More Information
Set Membership: |
|

ALDH5A1
Genomic Map