Assay Details
| Gene Symbol: FAM126B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194005.2 | NP_001180934.1 | 1023 | 138 | ||
| XM_015110620.2 | XP_014966106.1 | 1048 | 138 | |||
| XM_015110621.2 | XP_014966107.1 | 1102 | 138 | |||
| XM_015110622.2 | XP_014966108.1 | 922 | 138 | |||
| XM_015110623.2 | XP_014966109.1 | 967 | 138 | |||
| XM_015110624.2 | XP_014966110.1 | 1104 | 138 | |||
| XM_015110625.2 | XP_014966111.1 | 1194 | 138 | |||
| XM_028830460.1 | XP_028686293.1 | 1062 | 138 | |||
| XM_028830461.1 | XP_028686294.1 | 1103 | 138 | |||
| GenBank mRNA | JU334097.1 | - | 1017 | 138 | ||
| JU475244.1 | - | 989 | 138 | |||
More Information
Set Membership: |
|

FAM126B
Genomic Map