Assay Details
| Gene Symbol: MFSD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122459.2 | XP_014977945.2 | 1086 | 102 | ||
| XM_015122460.2 | XP_014977946.2 | 1086 | 102 | |||
| XM_028839226.1 | XP_028695059.1 | 1086 | 102 | |||
| XM_028839227.1 | XP_028695060.1 | 1086 | 102 | |||
| GenBank mRNA | JU325432.1 | - | 900 | 102 | ||
| JU474296.1 | - | 1055 | 102 | |||
| JV046178.1 | - | 1038 | 102 | |||
| JV667314.1 | - | 1044 | 102 | |||
More Information
Set Membership: |
|

MFSD12
Genomic Map