Assay Details
| Gene Symbol: COLEC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838240.1 | XP_028694073.1 | 2280 | 57 | ||
| GenBank mRNA | JU324960.1 | - | 393 | 57 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

COLEC12
Genomic Map