Assay Details
| Gene Symbol: ACVR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028843963.1 | XP_028699796.1 | 1365 | 61 | ||
| XM_028843964.1 | XP_028699797.1 | 1565 | 61 | |||
| GenBank mRNA | JU473069.1 | - | 1239 | 61 | ||
More Information
Set Membership: |
|

ACVR2B
Genomic Map