Assay Details
| Gene Symbol: MPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261546.1 | NP_001248475.1 | 976 | 84 | ||
| XM_015128494.2 | XP_014983980.1 | 965 | 84 | |||
| XM_028841745.1 | XP_028697578.1 | 1086 | 84 | |||
| XM_028841746.1 | XP_028697579.1 | 1017 | 84 | |||
| XM_028841747.1 | XP_028697580.1 | 1082 | 84 | |||
| GenBank mRNA | JU321530.1 | - | 993 | 84 | ||
| JU475661.1 | - | 976 | 84 | |||
| JV582359.1 | - | 1001 | 84 | |||
More Information
Set Membership: |
|

MPP1
Genomic Map