Assay Details
| Gene Symbol: TBXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101421.4 | XP_001101421.1 | 1283 | 132 | ||
| XM_015137661.2 | XP_014993147.2 | 1886 | 132 | |||
More Information
Set Membership: |
|

TBXT
Genomic Map