Assay Details
| Gene Symbol: DUSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257450.2 | NP_001244379.1 | 961 | 65 | ||
| GenBank mRNA | JU321828.1 | - | 962 | 65 | ||
| JU471240.1 | - | 874 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DUSP1
Genomic Map