Assay Details
| Gene Symbol: EPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257917.1 | NP_001244846.1 | 1063 | 119 | ||
| XM_015151109.2 | XP_015006595.1 | 1127 | 119 | |||
| XM_015151112.2 | XP_015006598.1 | 1097 | 119 | |||
| XM_015151114.2 | XP_015006600.1 | 1180 | 119 | |||
| XM_015151115.2 | XP_015006601.1 | 1109 | 119 | |||
| XM_028828853.1 | XP_028684686.1 | 1050 | 119 | |||
| GenBank mRNA | JU333351.1 | - | 1063 | 119 | ||
| JU333352.1 | - | 1063 | 119 | |||
| JV043990.1 | - | 1055 | 119 | |||
More Information
Set Membership: |
|

EPS8
Genomic Map