Assay Details
| Gene Symbol: SYT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265809.1 | NP_001252738.1 | 469 | 81 | ||
| XM_015118755.2 | XP_014974241.1 | 734 | 81 | |||
| XM_015118761.2 | XP_014974247.1 | 744 | 81 | |||
| XM_028843268.1 | XP_028699101.1 | 790 | 81 | |||
| XM_028843282.1 | XP_028699115.1 | 640 | 81 | |||
| GenBank mRNA | JV635777.1 | - | 469 | 81 | ||
More Information
Set Membership: |
|

SYT2
Genomic Map