Assay Details
| Gene Symbol: NLRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115010.2 | XP_014970496.2 | 2953 | 74 | ||
| XM_015115011.2 | XP_014970497.2 | 2953 | 74 | |||
| XM_028833744.1 | XP_028689577.1 | 3151 | 74 | |||
More Information
Set Membership: |
|

NLRP14
Genomic Map