Assay Details
| Gene Symbol: SLC13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271645.1 | NP_001258574.1 | 103 | 67 | ||
| GenBank mRNA | HM998307.1 | - | 103 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC13A5| Gene Symbol: SLC13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271645.1 | NP_001258574.1 | 103 | 67 | ||
| GenBank mRNA | HM998307.1 | - | 103 | 67 | ||
Set Membership: |
|