Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131199.2 | XP_014986685.2 | 1166 | 76 | ||
| XM_015131200.2 | XP_014986686.2 | 1071 | 76 | |||
| XM_015131201.2 | XP_014986687.2 | 3275 | 76 | |||
| XM_028844012.1 | XP_028699845.1 | 1263 | 76 | |||
| XM_028844013.1 | XP_028699846.1 | 1047 | 76 | |||
| XM_028844014.1 | XP_028699847.1 | 948 | 76 | |||
| XM_028844015.1 | XP_028699848.1 | 1035 | 76 | |||
| XM_028844016.1 | XP_028699849.1 | 365 | 76 | |||
| GenBank mRNA | JU334357.1 | - | 972 | 76 | ||
| JU475458.1 | - | 982 | 76 | |||
| JU475459.1 | - | 982 | 76 | |||
| JV046969.1 | - | 982 | 76 | |||
More Information
Set Membership: |
|

ZNF445
Genomic Map