Assay Details
| Gene Symbol: IMMP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086136.4 | XP_001086136.1 | 592 | 153 | ||
| XM_015114588.2 | XP_014970074.1 | 628 | 153 | |||
| XM_015114589.2 | XP_014970075.1 | 716 | 153 | |||
| XM_028833545.1 | XP_028689378.1 | 764 | 153 | |||
| XM_028833546.1 | XP_028689379.1 | 695 | 153 | |||
| GenBank mRNA | JU338575.1 | - | 723 | 153 | ||
| JV706583.1 | - | 734 | 153 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IMMP1L
Genomic Map