Assay Details
| Gene Symbol: ALAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101576.4 | XP_001101576.1 | 456 | 112 | ||
| XM_015117041.2 | XP_014972527.2 | 622 | 112 | |||
| GenBank mRNA | JU322586.1 | - | 399 | 112 | ||
| JU473124.1 | - | 398 | 112 | |||
| JV043813.1 | - | 447 | 112 | |||
| JV637382.1 | - | 453 | 112 | |||
More Information
Set Membership: |
|

ALAD
Genomic Map