Assay Details
| Gene Symbol: ACOT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266088.1 | NP_001253017.1 | 731 | 76 | ||
| XM_015148852.2 | XP_015004338.1 | 754 | 76 | |||
| XM_015148854.2 | XP_015004340.1 | 607 | 76 | |||
| GenBank mRNA | JU327233.1 | - | 1519 | 76 | ||
| JU472253.1 | - | 731 | 76 | |||
| JV597667.1 | - | 2033 | 76 | |||
More Information
Set Membership: |
|

ACOT8
Genomic Map