Assay Details
| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260704.1 | NP_001247633.1 | 996 | 94 | ||
| GenBank mRNA | JU336763.1 | - | 996 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM63| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260704.1 | NP_001247633.1 | 996 | 94 | ||
| GenBank mRNA | JU336763.1 | - | 996 | 94 | ||
Set Membership: |
|