Assay Details
| Gene Symbol: NONO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194913.2 | NP_001181842.1 | 1572 | 64 | ||
| XM_001091198.4 | XP_001091198.1 | 1787 | 64 | |||
| XM_001091316.4 | XP_001091316.1 | 1672 | 64 | |||
| XM_028841688.1 | XP_028697521.1 | 1150 | 64 | |||
| GenBank mRNA | JU320152.1 | - | 1291 | 64 | ||
| JV637384.1 | - | 1361 | 64 | |||
| JV668356.1 | - | 1274 | 64 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NONO
Genomic Map