Assay Details
| Gene Symbol: ZFAND5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117902.2 | XP_014973388.1 | 919 | 130 | ||
| XM_015117903.2 | XP_014973389.1 | 782 | 130 | |||
| GenBank mRNA | JU319734.1 | - | 937 | 130 | ||
| JU319735.1 | - | 800 | 130 | |||
| JU471540.1 | - | 931 | 130 | |||
| JU471541.1 | - | 794 | 130 | |||
| JV043184.1 | - | 968 | 130 | |||
| JV043185.1 | - | 831 | 130 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZFAND5
Genomic Map