Assay Details
| Gene Symbol: LRRC56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085909.4 | XP_001085909.2 | 2342 | 82 | ||
| XM_028832961.1 | XP_028688794.1 | 2469 | 82 | |||
| XM_028832962.1 | XP_028688795.1 | 2466 | 82 | |||
| XM_028832963.1 | XP_028688796.1 | 2339 | 82 | |||
| XM_028832964.1 | XP_028688797.1 | 2249 | 82 | |||
| XM_028832965.1 | XP_028688798.1 | 2342 | 82 | |||
More Information
Set Membership: |
|

LRRC56
Genomic Map