Assay Details
| Gene Symbol: LYPD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194767.3 | NP_001181696.1 | 368 | 132 | ||
| GenBank mRNA | JV045566.1 | - | 272 | 132 | ||
| JV635960.1 | - | 269 | 132 | |||
More Information
Set Membership: |
|

LYPD6
Genomic Map