Assay Details
| Gene Symbol: STK17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131912.2 | XP_014987398.1 | 939 | 67 | ||
| GenBank mRNA | JU322859.1 | - | 682 | 67 | ||
| JU477847.1 | - | 887 | 67 | |||
More Information
Set Membership: |
|

STK17B
Genomic Map