Assay Details
| Gene Symbol: GPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266539.1 | NP_001253468.1 | 975 | 109 | ||
| GenBank mRNA | JU322978.1 | - | 975 | 109 | ||
| JV044795.1 | - | 1065 | 109 | |||
| JV668102.1 | - | 1099 | 109 | |||
More Information
Set Membership: |
|

GPC1
Genomic Map