Assay Details
| Gene Symbol: CACNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266978.1 | NP_001253907.1 | 369 | 55 | ||
| XM_015151484.2 | XP_015006970.1 | 1039 | 55 | |||
| XM_015151485.2 | XP_015006971.1 | 320 | 55 | |||
| XM_028828911.1 | XP_028684744.1 | 244 | 55 | |||
| GenBank mRNA | JU320734.1 | - | 210 | 55 | ||
| JU477144.1 | - | 376 | 55 | |||
| JV048207.1 | - | 369 | 55 | |||
More Information
Set Membership: |
|

CACNB3
Genomic Map