Assay Details
| Gene Symbol: TMED7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204339.2 | NP_001191268.1 | 817 | 71 | ||
| GenBank mRNA | CK230008.1 | - | 205 | 71 | ||
| JU321744.1 | - | 738 | 71 | |||
| JU472466.1 | - | 624 | 71 | |||
| JV638144.1 | - | 688 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMED7
Genomic Map