Assay Details
| Gene Symbol: CENPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194334.2 | NP_001181263.1 | 203 | 111 | ||
| XM_028827317.1 | XP_028683150.1 | 231 | 111 | |||
| GenBank mRNA | JU328449.1 | - | 86 | 111 | ||
| JV593283.1 | - | 86 | 111 | |||
More Information
Set Membership: |
|

CENPM
Genomic Map