Assay Details
| Gene Symbol: BUD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266959.1 | NP_001253888.1 | 485 | 106 | ||
| XM_028845071.1 | XP_028700904.1 | 611 | 106 | |||
| GenBank mRNA | JU332790.1 | - | 491 | 106 | ||
| JU332791.1 | - | 491 | 106 | |||
| JU332792.1 | - | 491 | 106 | |||
| JU474210.1 | - | 491 | 106 | |||
| JV043191.1 | - | 485 | 106 | |||
| JV706194.1 | - | 507 | 106 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BUD23
Genomic Map