Assay Details
| Gene Symbol: NIF3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261733.1 | NP_001248662.1 | 462 | 74 | ||
| XM_015110614.2 | XP_014966100.1 | 846 | 74 | |||
| XM_015110615.2 | XP_014966101.1 | 869 | 74 | |||
| XM_015110616.2 | XP_014966102.1 | 725 | 74 | |||
| XM_028830455.1 | XP_028686288.1 | 847 | 74 | |||
| XM_028830456.1 | XP_028686289.1 | 845 | 74 | |||
| XM_028830457.1 | XP_028686290.1 | 845 | 74 | |||
| XM_028830458.1 | XP_028686291.1 | 845 | 74 | |||
| XM_028830459.1 | XP_028686292.1 | 843 | 74 | |||
| GenBank mRNA | JU335802.1 | - | 493 | 74 | ||
| JU476679.1 | - | 490 | 74 | |||
| JV047852.1 | - | 462 | 74 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NIF3L1
Genomic Map