Assay Details
| Gene Symbol: NIPSNAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150044.2 | XP_015005530.1 | 1059 | 66 | ||
| XM_028827864.1 | XP_028683697.1 | 1187 | 66 | |||
| GenBank mRNA | JU336820.1 | - | 633 | 66 | ||
| JV048507.1 | - | 644 | 66 | |||
| JV561016.1 | - | 478 | 66 | |||
| JV663996.1 | - | 649 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NIPSNAP1
Genomic Map