Assay Details
| Gene Symbol: NFS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097699.4 | XP_001097699.2 | 1490 | 65 | ||
| XM_015149110.2 | XP_015004596.1 | 1242 | 65 | |||
| XM_015149111.2 | XP_015004597.1 | 1221 | 65 | |||
| XM_015149112.2 | XP_015004598.1 | 1101 | 65 | |||
More Information
Set Membership: |
|

NFS1
Genomic Map