Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086177.4 | XP_001086177.3 | 544 | 139 | ||
| XM_015140607.2 | XP_014996093.1 | 280 | 139 | |||
| XM_015140608.2 | XP_014996094.1 | 213 | 139 | |||
| XM_015140610.2 | XP_014996096.1 | 252 | 139 | |||
| XM_015140611.2 | XP_014996097.1 | 597 | 139 | |||
| XM_015140612.2 | XP_014996098.1 | 596 | 139 | |||
| XM_028849668.1 | XP_028705501.1 | 280 | 139 | |||
| XM_028849669.1 | XP_028705502.1 | 280 | 139 | |||
| XM_028849670.1 | XP_028705503.1 | 280 | 139 | |||
| XM_028849671.1 | XP_028705504.1 | 252 | 139 | |||
| GenBank mRNA | JU319816.1 | - | 216 | 139 | ||
| JU474252.1 | - | 172 | 139 | |||
| JV046140.1 | - | 280 | 139 | |||
| JV641789.1 | - | 264 | 139 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AP3S1
Genomic Map